6-Chloro-2-methylnicotinonitrile
Lieferant: Apollo Scientific
OR310790-1G
APOSOR310790-1GEA
336
EUR
APOSOR310790-1G
6-Chloro-2-methylnicotinonitrile
6-Chloro-2-methylnicotinonitrile
Formel:
C₇H₅ClN₂ Molecular Weight: 152,581 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07082897 CAS-Nummer: 66909-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...