5-Chloro-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-6-azaindole

OR310792-5G
APOSOR310792-5GEA 75.4 EUR
APOSOR310792-5G
5-Chloro-1H-pyrrolo[2,3-c]pyridine
5-Chloro-1H-pyrrolo[2,3-c]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152.5800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09907859
CAS-Nummer: 131084-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR