1-(Cyclopropylmethyl)piperazine dihydrochloride

Lieferant: Apollo Scientific

OR310793-1G
APOSOR310793-1GEA 27.7 EUR
APOSOR310793-1G
1-(Cyclopropylmethyl)piperazine dihydrochloride
1-(Cyclopropylmethyl)piperazine dihydrochloride
Formel: C₈H₁₈Cl₂N₂
Molecular Weight: 213,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08544420
CAS-Nummer: 373608-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR