6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR310794-1G
APOSOR310794-1GEA
288
EUR
APOSOR310794-1G
6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine
6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine
Formel:
C₁₅H₁₅ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09040695 CAS-Nummer: 210539-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...