(2S)-1-Cyclobutyl-2-methylpiperazine dihydrochloride

Lieferant: Apollo Scientific

OR310796-500MG
APOSOR310796-500MGEA 560 EUR
APOSOR310796-500MG
(2S)-1-Cyclobutyl-2-methylpiperazine dihydrochloride
(2S)-1-Cyclobutyl-2-methylpiperazine dihydrochloride
Formel: C₉H₁₈N₂·2ClH
Molecular Weight: 227,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907868
CAS-Nummer: 1227610-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR