2-Methyl-5-phenylthiazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR310797-1G
APOSOR310797-1GEA 259 EUR
APOSOR310797-1G
2-Methyl-5-phenylthiazole-4-carboxylic acid
2-Methyl-5-phenylthiazole-4-carboxylic acid
Formel: C₁₁H₉NO₂S
Molecular Weight: 219,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04178838
CAS-Nummer: 13743-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR