(3-Aminopyrazin-2-yl)methanol

Lieferant: Apollo Scientific

OR310799-500MG
APOSOR310799-500MGEA 560 EUR
APOSOR310799-500MG
(3-Aminopyrazin-2-yl)methanol
(3-Aminopyrazin-2-yl)methanol
Formel: C₅H₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09907886
CAS-Nummer: 32710-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR