(4-Bromophenyl)methanesulphonamide

Lieferant: Apollo Scientific

OR310813-1G
APOSOR310813-1GEA 290 EUR
APOSOR310813-1G
(4-Bromophenyl)methanesulphonamide
(4-Bromophenyl)methanesulphonamide
Formel: C₇H₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12776301
CAS-Nummer: 64732-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR