5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane

Lieferant: Apollo Scientific

OR310870-1G OR310870-5G
APOSOR310870-1GEA 27.7 EUR
APOSOR310870-1G APOSOR310870-5G
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
Formel: C₁₂H₁₇BO₂
Molecular Weight: 204.073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101508
CAS-Nummer: 223799-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR