[3-(Methylamino)phenyl]methanol

Lieferant: Apollo Scientific

OR310887-500MG
APOSOR310887-500MGEA 782 EUR
APOSOR310887-500MG
[3-(Methylamino)phenyl]methanol
[3-(Methylamino)phenyl]methanol
Formel: C₈H₁₁NO
Molecular Weight: 137.179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658585
CAS-Nummer: 121562-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR