trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane
Lieferant: Apollo Scientific
|
Achtung
|
OR310891-250MG
OR310891-100MG
APOSOR310891-250MGEA
606
EUR
APOSOR310891-250MG
APOSOR310891-100MG
trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane
trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane
Formel:
C₆H₁₂O₄ Molecular Weight: 148.157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16659995 CAS-Nummer: 87133-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...