trans-2,5-Bis(iodomethyl)-1,4-dioxane

Lieferant: Apollo Scientific

OR310892-500MG
APOSOR310892-500MGEA 1350 EUR
APOSOR310892-500MG
trans-2,5-Bis(iodomethyl)-1,4-dioxane
trans-2,5-Bis(iodomethyl)-1,4-dioxane
Formel: C₆H₁₀I₂O₂
Molecular Weight: 367.951 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16659996
CAS-Nummer: 56127-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR