3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR310893-500MG
APOSOR310893-500MGEA
1350
EUR
APOSOR310893-500MG
3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
Formel:
C₈H₆N₂O₃ Molecular Weight: 178,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15526745 CAS-Nummer: 443956-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...