3-Chloro-8-nitroquinoline
Lieferant: Apollo Scientific
OR310904-500MG
APOSOR310904-500MGEA
782
EUR
APOSOR310904-500MG
3-Chloro-8-nitroquinoline
3-Chloro-8-nitroquinoline
Formel:
C₉H₅ClN₂O₂ Molecular Weight: 208,601 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044981 CAS-Nummer: 73868-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...