3-Chloroquinolin-8-amine

Lieferant: Apollo Scientific

OR310905-500MG
APOSOR310905-500MGEA 782 EUR
APOSOR310905-500MG
3-Chloroquinolin-8-amine
3-Chloroquinolin-8-amine
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040215
CAS-Nummer: 139399-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR