N-(4-Bromo-3-cyanophenyl)methanesulphonamide

Lieferant: Apollo Scientific

OR310907-500MG
APOSOR310907-500MGEA 661 EUR
APOSOR310907-500MG
N-(4-Bromo-3-cyanophenyl)methanesulphonamide
N-(4-Bromo-3-cyanophenyl)methanesulphonamide
Formel: C₈H₇BrN₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD16660030
CAS-Nummer: 1206776-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR