tert-Butyl 4-(pyridin-4-yloxy)piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR310911-1G OR310911-500MG
APOSOR310911-1GEA 894 EUR
APOSOR310911-1G APOSOR310911-500MG
tert-Butyl 4-(pyridin-4-yloxy)piperidine-1-carboxylate
tert-Butyl 4-(pyridin-4-yloxy)piperidine-1-carboxylate
Formel: C₁₅H₂₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09261175
CAS-Nummer: 308386-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR