1-Ethoxy-3-methoxybenzene

Lieferant: Apollo Scientific

OR310917-500MG
APOSOR310917-500MGEA 560 EUR
APOSOR310917-500MG
1-Ethoxy-3-methoxybenzene
1-Ethoxy-3-methoxybenzene
Formel: C₉H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21340356
CAS-Nummer: 25783-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR