Methyl 3-(4-methoxyphenyl)-3-methylbutanoate

Lieferant: Apollo Scientific

OR310919-500MG
APOSOR310919-500MGEA 1110 EUR
APOSOR310919-500MG
Methyl 3-(4-methoxyphenyl)-3-methylbutanoate
Methyl 3-(4-methoxyphenyl)-3-methylbutanoate
Formel: C₁₃H₁₈O₃
Molecular Weight: 222,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22688506
CAS-Nummer: 67437-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR