5-Amino-6-bromo-3,4-dihydro-2H-1-benzopyran-8-carboxylic acid

Lieferant: Apollo Scientific

OR310920-500MG
APOSOR310920-500MGEA 782 EUR
APOSOR310920-500MG
5-Amino-6-bromo-3,4-dihydro-2H-1-benzopyran-8-carboxylic acid
5-Amino-6-bromo-3,4-dihydro-2H-1-benzopyran-8-carboxylic acid
Formel: C₁₀H₁₀BrNO₃
Molecular Weight: 272,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17926267
CAS-Nummer: 941692-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR