tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate

OR310926-1G
APOSOR310926-1GEA 79.9 EUR
APOSOR310926-1G
tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate 95%
tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate
Formel: C₁₃H₁₇ClN₂O₂
Molecular Weight: 268.75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565958
CAS-Nummer: 1151665-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR