tert-Butyl 6-methoxy-7-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

Lieferant: Apollo Scientific

OR310935-500MG
APOSOR310935-500MGEA 1350 EUR
APOSOR310935-500MG
tert-Butyl 6-methoxy-7-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
tert-Butyl 6-methoxy-7-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
Formel: C₁₅H₂₀N₂O₅
Molecular Weight: 308,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101536
CAS-Nummer: 921224-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR