1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine

Lieferant: Apollo Scientific

OR310942-500MG
APOSOR310942-500MGEA 883 EUR
APOSOR310942-500MG
1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine
1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine
Formel: C₁₄H₂₅N₃O
Molecular Weight: 251,368 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261268
CAS-Nummer: 799559-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR