(6S)-6-(Propylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide

Lieferant: Apollo Scientific

OR310955-500MG
APOSOR310955-500MGEA 883 EUR
APOSOR310955-500MG
(6S)-6-(Propylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide
(6S)-6-(Propylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide
Formel: C₁₃H₁₉NO·BrH
Molecular Weight: 286,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20485837
CAS-Nummer: 165950-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR