5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate

Lieferant: Apollo Scientific

OR310958-500MG
APOSOR310958-500MGEA 1350 EUR
APOSOR310958-500MG
5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate
5-tert-butyl 3-ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3,5-dicarboxylate
Formel: C₁₄H₂₁N₃O₄
Molecular Weight: 295,334 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837793
CAS-Nummer: 1286753-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR