(4R)-3-(2-Bromoacetyl)-4-phenyl-1,3-oxazolidin-2-one

Lieferant: Apollo Scientific

OR310961-500MG
APOSOR310961-500MGEA 1350 EUR
APOSOR310961-500MG
(4R)-3-(2-Bromoacetyl)-4-phenyl-1,3-oxazolidin-2-one
(4R)-3-(2-Bromoacetyl)-4-phenyl-1,3-oxazolidin-2-one
Formel: C₁₁H₁₀BrNO₃
Molecular Weight: 284,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101542
CAS-Nummer: 227602-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR