2-{Spiro[2.2]pentan-1-yl}ethan-1-ol

Lieferant: Apollo Scientific

OR310965-500MG
APOSOR310965-500MGEA 1350 EUR
APOSOR310965-500MG
2-{Spiro[2.2]pentan-1-yl}ethan-1-ol
2-{Spiro[2.2]pentan-1-yl}ethan-1-ol
Formel: C₇H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28101543
CAS-Nummer: 118757-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR