(4-Vinylphenyl)boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

OR310973-1G OR310973-5G
APOSOR310973-1GEA 62.5 EUR
APOSOR310973-1G APOSOR310973-5G
(4-Vinylphenyl)boronic acid pinacol ester
(4-Vinylphenyl)boronic acid pinacol ester
Formel: C₁₄H₁₉BO₂
Lagertemperatur: Kühlschrank
MDL: MFCD18733870
CAS-Nummer: 870004-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR