Diethyl({[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine

Lieferant: Apollo Scientific

OR310980-1G OR310980-5G
APOSOR310980-1GEA 89.9 EUR
APOSOR310980-1G APOSOR310980-5G
Diethyl({[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine
Diethyl({[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine
Formel: C₁₇H₂₈BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09746204
CAS-Nummer: 1012785-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR