2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane

Lieferant: Apollo Scientific

OR310982-5G
APOSOR310982-5GEA 72.5 EUR
APOSOR310982-5G
2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
2-(4-Methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Formel: C₁₂H₁₇BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09909078
CAS-Nummer: 213596-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR