Ethyl 6-(chloromethyl)-4-(2-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

Lieferant: Apollo Scientific

OR310998-1G
APOSOR310998-1GEA 780 EUR
APOSOR310998-1G
Ethyl 6-(chloromethyl)-4-(2-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Ethyl 6-(chloromethyl)-4-(2-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Formel: C₁₄H₁₄Cl₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00749774
CAS-Nummer: 325479-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR