1,5-Dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-on
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Allopurinol
OR3110-25G
APOSOR3110-25GEA
0
EUR
APOSOR3110-25G
1,5-Dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-on
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
Formel:
C₅H₄N₄O Molecular Weight: 136,11 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00599413 CAS-Nummer: 315-30-0 EINECS: 206-250-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...