3-Anilino-5,5-dimethyl-2-cyclohexen-1-one

Lieferant: Apollo Scientific

OR311067-1G
APOSOR311067-1GEA 139 EUR
APOSOR311067-1G
3-Anilino-5,5-dimethyl-2-cyclohexen-1-one
3-Anilino-5,5-dimethyl-2-cyclohexen-1-one
Formel: C₁₄H₁₇NO
Molecular Weight: 215,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128479
CAS-Nummer: 18940-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR