3-(Acetylsulphanyl)-2-methylpropanamide
Lieferant: Apollo Scientific
OR311103-500MG
APOSOR311103-500MGEA
214
EUR
APOSOR311103-500MG
3-(Acetylsulphanyl)-2-methylpropanamide
3-(Acetylsulphanyl)-2-methylpropanamide
Formel:
C₆H₁₁NO₂S Molecular Weight: 161.222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02755772 CAS-Nummer: 909027-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...