3-Methoxy-4-(4-methyl-1H-imidazol-1-yl)benzoic acid

Lieferant: Apollo Scientific

OR311108-500MG
APOSOR311108-500MGEA 883 EUR
APOSOR311108-500MG
3-Methoxy-4-(4-methyl-1H-imidazol-1-yl)benzoic acid
3-Methoxy-4-(4-methyl-1H-imidazol-1-yl)benzoic acid
Formel: C₁₂H₁₂N₂O₃
Molecular Weight: 232,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18906996
CAS-Nummer: 937026-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR