4-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Lieferant: Apollo Scientific

Synonyme: 4-Nitro-1H-benzo[d]imidazol-2(3H)-one

OR311139-1G
APOSOR311139-1GEA 249 EUR
APOSOR311139-1G
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one
Formel: C₇H₅N₃O₃
Molecular Weight: 179,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835484
CAS-Nummer: 85330-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR