2-Chlorbenzamidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlorbenzoesäureamidin Hydrochlorid, 2-Chlorphenylamidin Hydrochlorid

OR311150-1G
APOSOR311150-1GEA 97.3 EUR
APOSOR311150-1G
2-Chlorbenzamidin Hydrochlorid
2-Chlorbenzamidin Hydrochlorid
Formel: C₇H₈Cl₂N₂
Molecular Weight: 191,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00275607
CAS-Nummer: 18637-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR