Cyclohex-3-ene-1-carbonyl chloride
Lieferant: Apollo Scientific
OR311203-1G
APOSOR311203-1GEA
174
EUR
APOSOR311203-1G
Cyclohex-3-ene-1-carbonyl chloride
Cyclohex-3-ene-1-carbonyl chloride
Formel:
C₇H₉ClO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013780 CAS-Nummer: 932-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...