3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-1-ol

Lieferant: Apollo Scientific

Synonyme: 3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-1-ol

OR311253-1G
APOSOR311253-1GEA 151 EUR
APOSOR311253-1G
3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-1-ol
3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-1-ol
Formel: C₁₅H₂₃BO₃
Molecular Weight: 262,152 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18734359
CAS-Nummer: 651030-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR