Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific

Synonyme: Allyl 3,5-diamino-1H-pyrazole-4-carboxylate, 3,5-diamino-1H-pyrazole-4-carboxylate

OR311292-500MG
APOSOR311292-500MGEA 336 EUR
APOSOR311292-500MG
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Formel: C₇H₁₀N₄O₂
Molecular Weight: 182,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27996731
CAS-Nummer: 1613191-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR