(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol

Lieferant: Apollo Scientific

OR311294-500MG
APOSOR311294-500MGEA 782 EUR
APOSOR311294-500MG
(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol
(1R)-1-(2-chlorophenyl)-2,2-dimethylpropane-1,3-diol
Formel: C₁₁H₁₅ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28976644
CAS-Nummer: 139406-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR