(1R)-1-(2-methoxyphenyl)-2,2-dimethylpropane-1,3-diol
Lieferant: Apollo Scientific
OR311296-500MG
APOSOR311296-500MGEA
782
EUR
APOSOR311296-500MG
(1R)-1-(2-methoxyphenyl)-2,2-dimethylpropane-1,3-diol
(1R)-1-(2-methoxyphenyl)-2,2-dimethylpropane-1,3-diol
Formel:
C₁₂H₁₈O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28976646 CAS-Nummer: 167255-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...