N-methoxy-N-methyl-1,2,3,6-tetrahydropyridine-2-carboxamide hydrochloride

Lieferant: Apollo Scientific

OR311298-500MG
APOSOR311298-500MGEA 782 EUR
APOSOR311298-500MG
N-methoxy-N-methyl-1,2,3,6-tetrahydropyridine-2-carboxamide hydrochloride
N-methoxy-N-methyl-1,2,3,6-tetrahydropyridine-2-carboxamide hydrochloride
Formel: C₈H₁₄N₂O₂·ClH
Molecular Weight: 206,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28976648
CAS-Nummer: 1823182-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR