2,2'-Dichloracetanilid
Lieferant: Apollo Scientific
OR3113-10G
OR3113-5G
APOSOR3113-10GEA
112
EUR
APOSOR3113-10G
APOSOR3113-5G
2,2'-Dichloracetanilid
2,2'-Dichloracetanilid
Formel:
C₈H₇Cl₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00032560 CAS-Nummer: 3289-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...