5-(Pyridin-2-yl)thiophene-2-sulphonohydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Hydrazinosulphonyl)-5-(pyridin-2-yl)thiophene, 2-[5-(Hydrazinosulphonyl)thien-2-yl]pyridine

OR3117-50MG
APOSOR3117-50MGEA 36.4 EUR
APOSOR3117-50MG
5-(Pyridin-2-yl)thiophene-2-sulphonohydrazide
5-(Pyridin-2-yl)thiophene-2-sulphonohydrazide
Formel: C₉H₉N₃O₂S₂
Molecular Weight: 255,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052124
CAS-Nummer: 175202-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR