4-(3-Hydroxyoxetan-3-yl)benzoic acid 95+%

Lieferant: Apollo Scientific

OR312032-250MG
APOSOR312032-250MGEA 95.7 EUR
APOSOR312032-250MG
4-(3-Hydroxyoxetan-3-yl)benzoic acid 95+%
4-(3-Hydroxyoxetan-3-yl)benzoic acid
Formel: C₁₀H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22370094
CAS-Nummer: 1346608-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR