1-Boc-3-ethynyl-3-hydroxyazetidine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-ethynyl-3-hydroxyazetidine-1-carboxylate
OR312034-250MG
APOSOR312034-250MGEA
171
EUR
APOSOR312034-250MG
1-Boc-3-ethynyl-3-hydroxyazetidine
1-Boc-3-ethynyl-3-hydroxyazetidine
Formel:
C₁₀H₁₅NO₃ Molecular Weight: 197,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21324699 CAS-Nummer: 1259034-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...