3-Aminooxetane-3-carboxylic acid, N-FMOC protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)oxetane-3-carboxylic acid, 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)oxetane-3-carboxylic acid

OR312038-1G
APOSOR312038-1GEA 719 EUR
APOSOR312038-1G
3-Aminooxetane-3-carboxylic acid, N-FMOC protected 97%
3-Aminooxetane-3-carboxylic acid, N-FMOC protected
Formel: C₁₉H₁₇NO₅
Molecular Weight: 339,342 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27664829
CAS-Nummer: 1380327-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR