1-Tosylazetidin-3-one
Lieferant: Apollo Scientific
OR312042-250MG
APOSOR312042-250MGEA
71.2
EUR
APOSOR312042-250MG
1-Tosylazetidin-3-one
1-Tosylazetidin-3-one
Formel:
C₁₀H₁₁NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129860 CAS-Nummer: 76543-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...