tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate

OR312050-250MG
APOSOR312050-250MGEA 445 EUR
APOSOR312050-250MG
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate
Formel: C₁₉H₂₈N₂O₄
Molecular Weight: 348,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22199157
CAS-Nummer: 1373923-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR